Search
Search

GLRX5| Gene Symbol: GLRX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016417.2 | NP_057501.2 | 1, 2 | 407 | ||
| GenBank mRNA | AB223038.1 | - | 1, 2 | 346 | ||
| BC023528.2 | - | 1, 2 | 357 | 3613760 | ||
| BC047680.1 | - | 1, 2 | 353 | 6066312 | ||
| BI259789.1 | - | 1, 2 | 508 | |||
| BI834047.1 | - | 1, 2 | 155 | |||
| BX248075.1 | - | 1, 2 | 360 | |||
| DQ083331.1 | - | 1, 2 | 303 | |||