Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKMY1 SIRNA Assays ›
Gene Symbol:
ANKMY1
Gene Name:
ankyrin repeat and MYND domain containing 1
Gene Aliases:
ZMYND13
Chromosome Location:
Chr.2: 240468632 - 240561110 on Build GRCh38
UniGene ID:
Hs.656615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135088
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKMY1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282771.1
NP_001269700.1
8
1953
NM_001282780.1
NP_001269709.1
6
1263
NM_001282781.1
NP_001269710.1
7
969
NM_001308375.1
NP_001295304.1
6
1502
NM_016552.3
NP_057636.2
7
1790
NM_017844.3
NP_060314.2
6
1263
XM_011511300.2
XP_011509602.1
8
2137
XM_011511301.1
XP_011509603.1
8
2137
XM_011511304.2
XP_011509606.1
8
2137
XM_011511305.2
XP_011509607.1
8
2137
XM_011511309.2
XP_011509611.1
8
2137
XM_011511313.2
XP_011509615.1
8
2137
XM_011511315.2
XP_011509617.1
8
2137
XM_011511317.1
XP_011509619.1
6
1447
XM_011511318.1
XP_011509620.1
6
1447
XM_011511319.2
XP_011509621.1
8
2137
XM_011511322.1
XP_011509624.1
6
1447
XM_017004271.1
XP_016859760.1
8
2137
XM_017004272.1
XP_016859761.1
8
2137
XM_017004273.1
XP_016859762.1
8
2137
XM_017004274.1
XP_016859763.1
8
2137
XM_017004275.1
XP_016859764.1
8
1903
XM_017004276.1
XP_016859765.1
9
2224
XM_017004277.1
XP_016859766.1
8
2137
XM_017004278.1
XP_016859767.1
8
2137
XM_017004279.1
XP_016859768.1
8
2137
XM_017004280.1
XP_016859769.1
8
2137
XM_017004281.1
XP_016859770.1
7
1743
XM_017004282.1
XP_016859771.1
7
1987
XM_017004283.1
XP_016859772.1
9
2224
XM_017004284.1
XP_016859773.1
6
1447
XM_017004285.1
XP_016859774.1
6
1213
XM_017004286.1
XP_016859775.1
7
1987
XM_017004287.1
XP_016859776.1
7
1987
XM_017004288.1
XP_016859777.1
5
1297
XM_017004289.1
XP_016859778.1
7
1987
XM_017004290.1
XP_016859779.1
6
1514
XM_017004291.1
XP_016859780.1
8
1957
GenBank mRNA
AB034636.1
-
7
1720
AK000506.1
-
6
1230
AK056511.2
-
3
987
AK124879.1
-
2
2312
AK125881.1
-
3
3272
AK126637.1
-
2
2764
AK225534.1
-
6
1230
AK296714.1
-
5
1016
AK314534.1
-
8
1937
AL136618.1
-
5
1009
AL833866.1
-
4
870
BC033495.1
-
6
1263
BC073146.1
-
6
1204
5743806
CR936600.1
-
6
1471
CR936759.1
-
6
1171
KU178642.1
-
5
1105
Back To Top
More Information
Gene Ontology Categories:
Process(es)
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0