Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016628.4 | NP_057712.2 | 13, 14 | 1965 | ||
NM_100264.2 | NP_567822.1 | 13, 14 | 2256 | |||
NM_100486.3 | NP_567823.1 | 12, 13 | 1656 | |||
XM_011519491.2 | XP_011517793.1 | 13, 14 | 2566 | |||
XM_017016315.1 | XP_016871804.1 | 13, 14 | 2073 | |||
XM_017016316.1 | XP_016871805.1 | 13, 14 | 2563 | |||
XM_017016317.1 | XP_016871806.1 | 12, 13 | 1767 | |||
XM_017016318.1 | XP_016871807.1 | 12, 13 | 1764 | |||
GenBank mRNA | AB058747.1 | - | 9, 10 | 4105 | ||
AF208858.1 | - | 6, 7 | 891 | |||
AK055852.1 | - | 13, 14 | 2073 | |||
AK091453.1 | - | 3, 4 | 2971 | |||
AK223600.1 | - | 13, 14 | 1963 | |||
AK225846.1 | - | 13, 14 | 2069 | |||
AK290174.1 | - | 13, 14 | 1970 | |||
AL713727.1 | - | 12, 13 | 2319 | |||
BC004258.1 | - | 13, 14 | 1956 | 3347345 | ||
BC010356.1 | - | 8, 9 | 1107 |