Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DISC1 SIRNA Assays ›
          Gene Symbol:
          DISC1 TSNAX-DISC1
          Gene Name:
          disrupted in schizophrenia 1
          TSNAX-DISC1 readthrough (NMD candidate)
          Gene Aliases:
          C1orf136, SCZD9
          -
          Chromosome Location:
          Chr.1: 231626815 - 232041272 on Build GRCh38
          Chromosome Location:
          Chr.1: 231528653 - 232041272 on Build GRCh38
          UniGene ID:
          Hs.13318
          Hs.13318
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 135225

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DISC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001012957.1 NP_001012975.1 2 925
          NM_001012958.1 NP_001012976.1 2 925
          NM_001012959.1 NP_001012977.1 2 925
          NM_001164537.1 NP_001158009.1 2 925
          NM_001164538.1 NP_001158010.1 2 925
          NM_001164539.1 NP_001158011.1 2 925
          NM_001164540.1 NP_001158012.1 2 925
          NM_001164541.1 NP_001158013.1 2 925
          NM_001164542.1 NP_001158014.1 2 925
          NM_001164544.1 NP_001158016.1 2 925
          NM_001164545.1 NP_001158017.1 2 925
          NM_001164546.1 NP_001158018.1 2 925
          NM_001164547.1 NP_001158019.1 2 925
          NM_001164548.1 NP_001158020.1 2 925
          NM_001164549.1 NP_001158021.1 2 925
          NM_001164550.1 NP_001158022.1 2 925
          NM_001164551.1 NP_001158023.1 2 925
          NM_001164552.1 NP_001158024.1 2 925
          NM_001164553.1 NP_001158025.1 2 925
          NM_001164554.1 NP_001158026.1 2 925
          NM_001164555.1 NP_001158027.1 2 925
          NM_018662.2 NP_061132.2 2 925
          GenBank mRNA AB007926.1 - 2 911
          AF222980.1 - 2 925
          AJ506177.1 - 2 925
          AJ506178.1 - 2 925
          AK025293.1 - 2 990
          AK096457.1 - 3 1097
          BC151225.1 - 2 911 9007180
          FJ804174.1 - 2 925
          FJ804175.1 - 2 925
          FJ804176.1 - 2 925
          FJ804177.1 - 2 925
          FJ804178.1 - 2 925
          FJ804179.1 - 2 925
          FJ804180.1 - 2 925
          FJ804181.1 - 2 925
          FJ804182.1 - 2 925
          FJ804183.1 - 2 925
          FJ804184.1 - 2 925
          FJ804185.1 - 2 925
          FJ804186.1 - 2 925
          FJ804187.1 - 2 925
          FJ804188.1 - 2 925
          FJ804189.1 - 2 925
          FJ804190.1 - 2 925
          FJ804191.1 - 2 925
          FJ804192.1 - 2 925
          FJ804193.1 - 2 925
          FJ804194.1 - 2 925
          FJ804195.1 - 3 993
          FJ804196.1 - 2 925
          FJ804197.1 - 2 925
          FJ804198.1 - 2 925
          FJ804199.1 - 2 925
          FJ804200.1 - 2 925
          FJ804201.1 - 2 925
          FJ804202.1 - 2 925
          FJ804203.1 - 2 925
          FJ804205.1 - 2 925
          FJ804206.1 - 2 925
          FJ804207.1 - 2 925
          FJ804208.1 - 2 925
          FJ804209.1 - 2 925
          FJ804210.1 - 2 925
          FJ804211.1 - 2 925
          FJ804212.1 - 2 925
          FJ804213.1 - 2 925
          FJ804214.1 - 2 925
          FJ804215.1 - 2 925
          FJ804216.1 - 2 925
          Gene Symbol: TSNAX-DISC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_028393.1 6 1593
          NR_028394.1 7 1721
          NR_028395.1 7 1721
          NR_028396.1 6 1593
          NR_028397.1 6 1458
          NR_028398.1 4 1212
          NR_028399.1 8 1828
          NR_028400.1 7 1700

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          neuron migration
          positive regulation of cell-matrix adhesion
          positive regulation of neuroblast proliferation
          cell proliferation in forebrain
          pyramidal neuron migration
          positive regulation of Wnt signaling pathway
          TOR signaling
          nonmotile primary cilium assembly
          positive regulation of axon extension
          mitochondrial calcium ion homeostasis
          response to electrical stimulus
          canonical Wnt signaling pathway
          regulation of dendritic spine development
          protein localization to centrosome
          regulation of synapse maturation
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          protein binding
          kinesin binding
          protein complex binding
          protein complex scaffold

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0