Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019850.2 | NP_062824.2 | 2 | 338 | ||
XM_011510923.2 | XP_011509225.1 | 2 | 304 | |||
GenBank mRNA | AK298315.1 | - | 2 | 290 | ||
BC031573.1 | - | 2 | 325 | 5186132 | ||
DB242442.1 | - | 2 | 338 |