Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291501.1 | NP_001278430.1 | 14, 15 | 2373 | ||
NM_148674.4 | NP_683515.4 | 14, 15 | 2373 | |||
XM_011530144.2 | XP_011528446.1 | 14, 15 | 2234 | |||
XM_011530145.2 | XP_011528447.1 | 14, 15 | 2425 | |||
GenBank mRNA | AJ504806.2 | - | 14, 15 | 2373 | ||
AK125736.1 | - | 1, 2 | 132 | |||
BC126208.1 | - | 14, 15 | 2353 | 8991924 |