Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024021.3 | NP_076926.2 | 6 | 728 | ||
NM_148975.2 | NP_683876.1 | 5 | 582 | |||
XM_017017909.1 | XP_016873398.1 | 5 | 595 | |||
GenBank mRNA | AB013102.1 | - | 5 | 423 | ||
AB022821.1 | - | 5 | 447 | |||
AF068288.1 | - | 5 | 516 | |||
AF237912.1 | - | 5 | 533 | |||
AF350500.1 | - | 4 | 486 | |||
AK075081.1 | - | 6 | 613 | |||
AK292089.1 | - | 5 | 537 | |||
AK314033.1 | - | 5 | 425 | |||
BC020648.1 | - | 5 | 457 | 4720088 | ||
BG538851.1 | - | 5 | 584 |