Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF3C SIRNA Assays ›
          Gene Symbol:
          EIF3C EIF3CL
          Gene Name:
          eukaryotic translation initiation factor 3 subunit C
          eukaryotic translation initiation factor 3 subunit C-like
          Gene Aliases:
          EIF3CL, EIF3S8, eIF3-p110
          -
          Chromosome Location:
          Chr.16: 28688558 - 28735730 on Build GRCh38
          Chromosome Location:
          Chr.16: 28379581 - 28403885 on Build GRCh38
          UniGene ID:
          Hs.567374
          Hs.567374
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 13544

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EIF3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037808.2 NP_001032897.1 2, 3 274
          NM_001199142.1 NP_001186071.1 2, 3 394
          NM_001267574.2 NP_001254503.1 1, 2 363
          NM_001286478.1 NP_001273407.1 2, 3 225
          NM_003752.4 NP_003743.1 2, 3 225
          XM_017023814.1 XP_016879303.1 2, 3 180
          GenBank mRNA AK000739.1 - 1, 2 255
          AK055366.1 - 2, 3 168
          AK292155.1 - 2, 3 394
          AK293264.1 - 2, 3 166
          AK299421.1 - 2, 3 168
          AK301183.1 - 2, 3 178
          AK301233.1 - 2, 3 177
          AK303790.1 - 2, 3 168
          AK304500.1 - 2, 3 170
          AL045958.1 - 2, 3 177
          AY762099.1 - 2, 3 137
          BC000533.2 - 2, 3 155
          BC001571.1 - 2, 3 159 3452953
          BC071705.1 - 2, 3 153 5552014
          BC107692.1 - 2, 3 185
          BC157842.1 - 2, 3 283 9057061
          BC157849.1 - 1, 2 123 9057068
          BE780094.1 - 2, 3 203
          BF665714.1 - 2, 3 182
          BM757468.1 - 2, 3 179
          DC341554.1 - 2, 3 225
          Gene Symbol: EIF3CL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099661.2 NP_001093131.1 2, 3 226
          NM_001317856.1 NP_001304785.1 2, 3 271
          NM_001317857.1 NP_001304786.1 2, 3 222
          XM_017023620.1 XP_016879109.1 2, 3 178
          GenBank mRNA BQ686089.1 - 2, 3 191
          DA645577.1 - 2, 3 172
          DC410988.1 - 2, 3 222

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          formation of translation preinitiation complex
          translational initiation
          regulation of translational initiation
          translation initiation factor activity
          protein binding
          translation initiation factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0