Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNIP1 SIRNA Assays ›
Gene Symbol:
TNIP1
Gene Name:
TNFAIP3 interacting protein 1
Gene Aliases:
ABIN-1, NAF1, VAN, nip40-1
Chromosome Location:
Chr.5: 151029943 - 151087660 on Build GRCh38
UniGene ID:
Hs.355141
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135563
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252385.1
NP_001239314.1
3, 4
520
NM_001252386.1
NP_001239315.1
2, 3
348
NM_001252390.1
NP_001239319.1
3, 4
385
NM_001252391.1
NP_001239320.1
3, 4
417
NM_001252392.1
NP_001239321.1
3, 4
417
NM_001252393.1
NP_001239322.1
3, 4
520
NM_001258454.1
NP_001245383.1
3, 4
520
NM_001258455.1
NP_001245384.1
2, 3
315
NM_001258456.1
NP_001245385.1
2, 3
315
NM_006058.4
NP_006049.3
3, 4
520
XM_005268355.1
XP_005268412.1
3, 4
433
XM_006714751.1
XP_006714814.1
3, 4
433
XM_006714752.2
XP_006714815.1
3, 4
433
XM_017008945.1
XP_016864434.1
3, 4
433
XM_017008946.1
XP_016864435.1
3, 4
399
XM_017008947.1
XP_016864436.1
4, 5
902
XM_017008948.1
XP_016864437.1
2, 3
694
XM_017008949.1
XP_016864438.1
3, 4
433
XM_017008950.1
XP_016864439.1
3, 4
433
XM_017008951.1
XP_016864440.1
2, 3
694
GenBank mRNA
AB177543.1
-
2, 3
279
AB177544.1
-
2, 3
279
AB252970.1
-
3, 4
389
AB252971.1
-
3, 4
389
AB252972.1
-
3, 4
510
AB252973.1
-
3, 4
510
AB252976.1
-
1, 2
143
AB252977.1
-
1, 2
143
AB252978.1
-
1, 2
143
AB252979.1
-
1, 2
143
AB252980.1
-
1, 2
143
AB252981.1
-
1, 2
143
AB252982.1
-
3, 4
389
AJ011895.1
-
3, 4
389
AJ011896.1
-
3, 4
389
AK074400.1
-
3, 4
451
AK290999.1
-
3, 4
386
AK298685.1
-
2, 3
342
AK299975.1
-
2, 3
280
AK303573.1
-
3, 4
470
AY012155.1
-
3, 4
536
BC012133.1
-
3, 4
426
4549542
BC014008.1
-
3, 4
367
4040118
BP254790.1
-
3, 4
510
BX640647.1
-
2, 3
269
D30755.2
-
2, 3
279
DA643531.1
-
2, 3
245
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MyD88-dependent toll-like receptor signaling pathway
translation
proteolysis
defense response
inflammatory response
leukocyte cell-cell adhesion
glycoprotein biosynthetic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of viral genome replication
positive regulation of transcription from RNA polymerase II promoter
positive regulation of inflammatory response
negative regulation of ERK1 and ERK2 cascade
modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
thiol-dependent ubiquitin-specific protease activity
protein binding
mitogen-activated protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0