Search
Search

TUBGCP3| Gene Symbol: TUBGCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286277.1 | NP_001273206.1 | 22 | 3689 | ||
| NM_006322.5 | NP_006313.1 | 22 | 3719 | |||
| XM_011537462.2 | XP_011535764.1 | 20 | 3372 | |||
| XM_011537463.2 | XP_011535765.1 | 23 | 3648 | |||
| XM_011537464.2 | XP_011535766.1 | 22 | 4452 | |||
| XM_017020323.1 | XP_016875812.1 | 23 | 4066 | |||
| GenBank mRNA | AF042378.1 | - | 22 | 3598 | ||
| BC046634.2 | - | 22 | 3702 | 5295831 | ||
| L13800.1 | - | 1 | 258 | |||