Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006420.2 | NP_006411.2 | 6 | 884 | ||
XM_005260252.3 | XP_005260309.1 | 6 | 901 | |||
GenBank mRNA | AF084521.1 | - | 6 | 732 | ||
BC050449.1 | - | 6 | 884 | |||
BC144625.1 | - | 6 | 795 | |||
BC144626.1 | - | 6 | 795 |