Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135876
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
14
4736
NM_001025077.2
NP_001020248.1
13
5933
NM_001083591.1
NP_001077060.1
14
4730
NM_006561.3
NP_006552.3
14
4682
XM_005252354.4
XP_005252411.1
14
4769
XM_005252357.4
XP_005252414.1
11
5728
XM_005252358.3
XP_005252415.1
12
4711
XM_006717373.2
XP_006717436.1
13
4809
XM_011519294.2
XP_011517596.1
14
4751
XM_011519297.1
XP_011517599.1
13
6232
XM_017015542.1
XP_016871031.1
13
4457
XM_017015543.1
XP_016871032.1
13
5905
XM_017015544.1
XP_016871033.1
13
5893
XM_017015545.1
XP_016871034.1
13
5887
XM_017015546.1
XP_016871035.1
14
4529
XM_017015547.1
XP_016871036.1
13
6139
XM_017015548.1
XP_016871037.1
13
5875
XM_017015549.1
XP_016871038.1
14
4517
XM_017015550.1
XP_016871039.1
13
6873
XM_017015551.1
XP_016871040.1
14
4763
XM_017015552.1
XP_016871041.1
13
6262
XM_017015553.1
XP_016871042.1
13
6250
XM_017015554.1
XP_016871043.1
14
4874
XM_017015556.1
XP_016871045.1
12
6179
XM_017015557.1
XP_016871046.1
14
5844
XM_017015558.1
XP_016871047.1
12
5831
XM_017015559.1
XP_016871048.1
12
6167
XM_017015560.1
XP_016871049.1
12
5828
XM_017015561.1
XP_016871050.1
12
6155
XM_017015562.1
XP_016871051.1
12
6149
XM_017015563.1
XP_016871052.1
12
6136
XM_017015564.1
XP_016871053.1
11
5755
XM_017015565.1
XP_016871054.1
11
5758
XM_017015566.1
XP_016871055.1
11
5743
XM_017015567.1
XP_016871056.1
12
5883
XM_017015568.1
XP_016871057.1
14
5870
XM_017015569.1
XP_016871058.1
13
4806
XM_017015570.1
XP_016871059.1
11
5740
XM_017015571.1
XP_016871060.1
12
6152
XM_017015572.1
XP_016871061.1
13
4495
GenBank mRNA
AF036956.1
-
14
4657
AF090693.1
-
13
5914
AF090694.1
-
14
4525
CR936676.1
-
2
6134
DA310511.1
-
1
440
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0