Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303618.1 | NP_001290547.1 | 4 | 1189 | ||
NM_001303619.1 | NP_001290548.1 | 3 | 782 | |||
NM_006566.3 | NP_006557.2 | 5 | 1191 | |||
XM_005266642.3 | XP_005266699.1 | 5 | 1290 | |||
XM_005266643.3 | XP_005266700.1 | 3 | 384 | |||
XM_006722374.3 | XP_006722437.1 | 3 | 855 | |||
XM_017025524.1 | XP_016881013.1 | 5 | 7934 | |||
XM_017025525.1 | XP_016881014.1 | 3 | 666 | |||
XM_017025526.1 | XP_016881015.1 | 4 | 560 | |||
XM_017025527.1 | XP_016881016.1 | 3 | 1224 | |||
GenBank mRNA | AJ537553.1 | - | 4 | 771 | ||
AK313199.1 | - | 4 | 1190 | |||
BC074787.2 | - | 4 | 776 | 30915562 | ||
GU935335.1 | - | 5 | 938 | |||
GU935336.1 | - | 3 | 541 | |||
U56102.1 | - | 5 | 950 |