Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006710.4 | NP_006701.1 | 5 | 902 | ||
NM_198189.2 | NP_937832.1 | 6 | 967 | |||
GenBank mRNA | AK289891.1 | - | 6 | 501 | ||
AK294078.1 | - | 5 | 884 | |||
AK308962.1 | - | 5 | 856 | |||
AK313437.1 | - | 5 | 445 | |||
BC003090.1 | - | 5 | 426 | 3503287 | ||
BC036499.1 | - | 5 | 1036 | 5264527 | ||
BC080617.1 | - | 5 | 398 | 6252628 | ||
BF979628.1 | - | 531 | ||||
BG721393.1 | - | 6 | 505 | |||
CR456994.1 | - | 5 | 378 | |||
U51205.1 | - | 5 | 427 |