Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006809.4 | NP_006800.2 | 7 | 1006 | ||
XM_005260254.2 | XP_005260311.1 | 7 | 939 | |||
XM_011528501.1 | XP_011526803.1 | 7 | 825 | |||
XM_017027600.1 | XP_016883089.1 | 5 | 1144 | |||
GenBank mRNA | AB085681.1 | - | 7 | 931 | ||
AK000594.1 | - | 7 | 925 | |||
AK129538.1 | - | 6 | 507 | |||
AK222952.1 | - | 7 | 913 | |||
AK302128.1 | - | 7 | 835 | |||
AK307655.1 | - | 7 | 947 | |||
BC001763.1 | - | 7 | 919 | 3544124 | ||
BC007423.2 | - | 7 | 919 | 2960090 | ||
BC014907.1 | - | 7 | 896 | 3922502 | ||
BT020008.1 | - | 7 | 866 | |||
BT020009.1 | - | 7 | 866 | |||
CR456765.1 | - | 7 | 866 | |||
U58970.1 | - | 7 | 897 |