Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS SIRNA Assays ›
Gene Symbol:
CADPS
Gene Name:
calcium dependent secretion activator
Gene Aliases:
CADPS1, CAPS, CAPS1, UNC-31
Chromosome Location:
Chr.3: 62398346 - 62875389 on Build GRCh38
UniGene ID:
Hs.654933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13614
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003716.3
NP_003707.2
26
3972
NM_183393.2
NP_899630.1
23
3735
NM_183394.2
NP_899631.1
24
3855
XM_006713378.2
XP_006713441.2
24
3793
XM_011534177.1
XP_011532479.1
29
4201
XM_011534178.1
XP_011532480.1
28
4183
XM_011534180.1
XP_011532482.1
29
4153
XM_011534182.1
XP_011532484.1
28
4132
XM_011534183.1
XP_011532485.1
27
4114
XM_011534189.1
XP_011532491.1
27
4036
XM_011534191.2
XP_011532493.1
25
3642
XM_011534192.1
XP_011532494.1
28
3994
XM_011534193.1
XP_011532495.1
28
3982
XM_011534194.2
XP_011532496.1
27
3624
XM_011534195.1
XP_011532497.1
26
3967
XM_011534196.1
XP_011532498.1
27
3964
XM_011534197.2
XP_011532499.1
24
3898
XM_011534198.2
XP_011532500.1
24
3444
XM_011534199.2
XP_011532501.1
24
3441
XM_011534200.2
XP_011532502.1
23
3426
XM_011534203.1
XP_011532505.1
26
3949
XM_017007358.1
XP_016862847.1
29
4207
XM_017007359.1
XP_016862848.1
29
4195
XM_017007360.1
XP_016862849.1
28
4186
XM_017007361.1
XP_016862850.1
28
4138
XM_017007362.1
XP_016862851.1
28
4126
XM_017007363.1
XP_016862852.1
27
4117
XM_017007364.1
XP_016862853.1
27
4039
XM_017007365.1
XP_016862854.1
27
4027
XM_017007366.1
XP_016862855.1
26
4021
XM_017007367.1
XP_016862856.1
26
3660
XM_017007368.1
XP_016862857.1
26
3657
XM_017007369.1
XP_016862858.1
28
3994
XM_017007370.1
XP_016862859.1
27
3979
XM_017007371.1
XP_016862860.1
26
3970
XM_017007372.1
XP_016862861.1
27
3615
XM_017007373.1
XP_016862862.1
27
3967
XM_017007374.1
XP_016862863.1
26
3958
XM_017007375.1
XP_016862864.1
26
3600
XM_017007376.1
XP_016862865.1
25
3952
XM_017007377.1
XP_016862866.1
25
3591
XM_017007378.1
XP_016862867.1
25
3588
XM_017007379.1
XP_016862868.1
25
3940
XM_017007380.1
XP_016862869.1
26
3883
XM_017007381.1
XP_016862870.1
26
3880
XM_017007382.1
XP_016862871.1
25
3513
XM_017007383.1
XP_016862872.1
25
3862
XM_017007384.1
XP_016862873.1
26
3832
XM_017007385.1
XP_016862874.1
26
3820
XM_017007386.1
XP_016862875.1
25
3811
GenBank mRNA
AB032947.2
-
26
3962
AB593079.1
-
24
3898
AF458662.1
-
23
3685
AL833895.1
-
24
3152
BC015754.1
-
4
303
BC072684.1
-
3
253
BC131731.1
-
17
1930
40124561
BX647166.1
-
6
587
U36448.1
-
4
321
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
protein transport
vesicle organization
synaptic vesicle priming
catecholamine secretion
dense core granule exocytosis
protein binding
lipid binding
protein kinase binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0