Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NT5C2 SIRNA Assays ›
          Gene Symbol:
          NT5C2
          Gene Name:
          5'-nucleotidase, cytosolic II
          Gene Aliases:
          GMP, NT5B, PNT5, SPG45, SPG65, cN-II
          Chromosome Location:
          Chr.10: 103088017 - 103193306 on Build GRCh38
          UniGene ID:
          Hs.97439
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136451

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NT5C2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134373.2 NP_001127845.1 6, 7 577
          NM_012229.4 NP_036361.1 7, 8 675
          XM_005269632.4 XP_005269689.1 7, 8 586
          XM_005269633.4 XP_005269690.1 8, 9 679
          XM_005269634.4 XP_005269691.1 8, 9 711
          XM_005269635.4 XP_005269692.1 7, 8 901
          XM_005269636.4 XP_005269693.1 7, 8 687
          XM_005269637.4 XP_005269694.1 6, 7 1405
          XM_005269638.4 XP_005269695.1 6, 7 2706
          XM_005269639.4 XP_005269696.1 5, 6 430
          XM_005269640.4 XP_005269697.1 8, 9 702
          XM_005269641.4 XP_005269698.1 7, 8 604
          XM_005269642.4 XP_005269699.1 6, 7 529
          XM_005269643.4 XP_005269700.1 5, 6 469
          XM_005269644.4 XP_005269701.1 9, 10 726
          XM_005269645.4 XP_005269702.1 8, 9 628
          XM_005269646.4 XP_005269703.1 7, 8 554
          XM_006717721.3 XP_006717784.1 7, 8 628
          XM_006717723.3 XP_006717786.1 5, 6 425
          XM_011539537.2 XP_011537839.1 8, 9 711
          XM_017015947.1 XP_016871436.1 5, 6 455
          XM_017015948.1 XP_016871437.1 8, 9 811
          XM_017015949.1 XP_016871438.1 7, 8 604
          XM_017015952.1 XP_016871441.1 6, 7 543
          XM_017015953.1 XP_016871442.1 7, 8 938
          XM_017015954.1 XP_016871443.1 6, 7 877
          XM_017015955.1 XP_016871444.1 6, 7 543
          XM_017015956.1 XP_016871445.1 6, 7 877
          XM_017015957.1 XP_016871446.1 8, 9 628
          XM_017015958.1 XP_016871447.1 9, 10 726
          XM_017015959.1 XP_016871448.1 6, 7 530
          XM_017015960.1 XP_016871449.1 7, 8 493
          XM_017015961.1 XP_016871450.1 7, 8 888
          XM_017015962.1 XP_016871451.1 6, 7 530
          XM_017015963.1 XP_016871452.1 6, 7 613
          XM_017015964.1 XP_016871453.1 6, 7 613
          XM_017015965.1 XP_016871454.1 5, 6 490
          XM_017015966.1 XP_016871455.1 5, 6 469
          XM_017015967.1 XP_016871456.1 5, 6 425
          XM_017015972.1 XP_016871461.1 7, 8 604
          XM_017015973.1 XP_016871462.1 8, 9 628
          XM_017015974.1 XP_016871463.1 7, 8 628
          XM_017015975.1 XP_016871464.1 5, 6 469
          GenBank mRNA AK127670.1 - 6, 7 385
          AK291667.1 - 6, 7 543
          AK295593.1 - 5, 6 719
          AK312517.1 - 6, 7 543
          BC001595.1 - 6, 7 626 3543103
          BX648078.1 - 8, 9 708
          D38524.1 - 6, 7 565

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotide phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide catabolic process
          phosphorylation
          dephosphorylation
          drug metabolic process
          IMP metabolic process
          adenosine metabolic process
          nucleotide binding
          protein binding
          5'-nucleotidase activity
          metal ion binding
          nucleoside phosphotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0