Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NT5C2 SIRNA Assays ›
          Gene Symbol:
          NT5C2
          Gene Name:
          5'-nucleotidase, cytosolic II
          Gene Aliases:
          GMP, NT5B, PNT5, SPG45, SPG65, cN-II
          Chromosome Location:
          Chr.10: 103088017 - 103193306 on Build GRCh38
          UniGene ID:
          Hs.97439
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136452

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NT5C2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134373.2 NP_001127845.1 8, 9 736
          NM_012229.4 NP_036361.1 9, 10 834
          XM_005269632.4 XP_005269689.1 9, 10 745
          XM_005269633.4 XP_005269690.1 10, 11 838
          XM_005269634.4 XP_005269691.1 10, 11 870
          XM_005269635.4 XP_005269692.1 9, 10 1060
          XM_005269636.4 XP_005269693.1 9, 10 846
          XM_005269637.4 XP_005269694.1 8, 9 1564
          XM_005269638.4 XP_005269695.1 8, 9 2865
          XM_005269639.4 XP_005269696.1 7, 8 589
          XM_005269640.4 XP_005269697.1 10, 11 861
          XM_005269641.4 XP_005269698.1 9, 10 763
          XM_005269642.4 XP_005269699.1 8, 9 688
          XM_005269643.4 XP_005269700.1 7, 8 628
          XM_005269644.4 XP_005269701.1 11, 12 885
          XM_005269645.4 XP_005269702.1 10, 11 787
          XM_005269646.4 XP_005269703.1 9, 10 713
          XM_006717721.3 XP_006717784.1 9, 10 787
          XM_006717723.3 XP_006717786.1 7, 8 584
          XM_011539537.2 XP_011537839.1 10, 11 870
          XM_017015947.1 XP_016871436.1 7, 8 614
          XM_017015948.1 XP_016871437.1 10, 11 970
          XM_017015949.1 XP_016871438.1 10, 11 955
          XM_017015950.1 XP_016871439.1 8, 9 671
          XM_017015951.1 XP_016871440.1 7, 8 597
          XM_017015952.1 XP_016871441.1 9, 10 894
          XM_017015953.1 XP_016871442.1 10, 11 1289
          XM_017015954.1 XP_016871443.1 9, 10 1228
          XM_017015955.1 XP_016871444.1 9, 10 915
          XM_017015956.1 XP_016871445.1 9, 10 1121
          XM_017015957.1 XP_016871446.1 11, 12 979
          XM_017015958.1 XP_016871447.1 12, 13 1098
          XM_017015959.1 XP_016871448.1 8, 9 689
          XM_017015960.1 XP_016871449.1 9, 10 652
          XM_017015961.1 XP_016871450.1 9, 10 1047
          XM_017015962.1 XP_016871451.1 9, 10 881
          XM_017015963.1 XP_016871452.1 9, 10 985
          XM_017015964.1 XP_016871453.1 9, 10 964
          XM_017015965.1 XP_016871454.1 8, 9 841
          XM_017015966.1 XP_016871455.1 8, 9 841
          XM_017015967.1 XP_016871456.1 8, 9 776
          XM_017015968.1 XP_016871457.1 9, 10 769
          XM_017015969.1 XP_016871458.1 9, 10 863
          XM_017015970.1 XP_016871459.1 7, 8 1506
          XM_017015971.1 XP_016871460.1 6, 7 550
          XM_017015972.1 XP_016871461.1 9, 10 763
          XM_017015973.1 XP_016871462.1 10, 11 787
          XM_017015974.1 XP_016871463.1 9, 10 787
          XM_017015975.1 XP_016871464.1 7, 8 628
          XM_017015976.1 XP_016871465.1 7, 8 634
          GenBank mRNA AK127670.1 - 9, 10 825
          AK291667.1 - 8, 9 702
          AK295593.1 - 7, 8 878
          AK312517.1 - 8, 9 702
          BC001595.1 - 8, 9 785 3543103
          BX648078.1 - 11, 12 1080
          D38524.1 - 8, 9 724

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          nucleotide phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide catabolic process
          phosphorylation
          dephosphorylation
          drug metabolic process
          IMP metabolic process
          adenosine metabolic process
          nucleotide binding
          protein binding
          5'-nucleotidase activity
          metal ion binding
          nucleoside phosphotransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0