Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012311.3 | NP_036443.1 | 7 | 713 | ||
NR_045609.1 | 7 | 713 | ||||
XM_006717434.3 | XP_006717497.1 | 7 | 708 | |||
XM_006717435.3 | XP_006717498.1 | 6 | 581 | |||
XM_011519427.1 | XP_011517729.1 | 7 | 708 | |||
XM_017015946.1 | XP_016871435.1 | 6 | 584 | |||
GenBank mRNA | AJ005273.1 | - | 7 | 684 | ||
AK296637.1 | - | 6 | 581 | |||
AK301789.1 | - | 7 | 713 | |||
AK309059.1 | - | 7 | 649 | |||
BC017309.2 | - | 7 | 630 | 5089578 |