Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS1 SIRNA Assays ›
Gene Symbol:
RIMS1
Gene Name:
regulating synaptic membrane exocytosis 1
Gene Aliases:
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location:
Chr.6: 71886703 - 72403143 on Build GRCh38
UniGene ID:
Hs.485729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136690
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
FAM | 20 nmol
100 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014989.5
NP_055804.2
5
957
XM_005248688.3
XP_005248745.1
5
893
XM_011535604.2
XP_011533906.2
5
893
XM_011535611.2
XP_011533913.2
5
893
XM_011535615.2
XP_011533917.2
5
893
XM_017010516.1
XP_016866005.1
5
893
XM_017010517.1
XP_016866006.1
5
893
XM_017010518.1
XP_016866007.1
5
893
XM_017010519.1
XP_016866008.1
5
893
XM_017010520.1
XP_016866009.1
5
893
XM_017010521.1
XP_016866010.1
5
893
XM_017010522.1
XP_016866011.1
5
893
XM_017010523.1
XP_016866012.1
5
893
XM_017010524.1
XP_016866013.1
5
893
XM_017010525.1
XP_016866014.1
5
893
XM_017010526.1
XP_016866015.1
5
893
XM_017010527.1
XP_016866016.1
5
893
XM_017010528.1
XP_016866017.1
5
893
XM_017010529.1
XP_016866018.1
5
893
XM_017010530.1
XP_016866019.1
5
893
XM_017010531.1
XP_016866020.1
5
893
XM_017010532.1
XP_016866021.1
5
893
XM_017010533.1
XP_016866022.1
5
893
GenBank mRNA
AB002338.1
-
5
713
AB045726.1
-
5
956
AB051866.1
-
5
956
AY190519.1
-
5
636
BC151853.1
-
5
737
8860502
BC152435.1
-
5
713
8862556
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
intracellular protein transport
neurotransmitter secretion
visual perception
positive regulation of gene expression
glutamate secretion
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulated exocytosis
secretion
regulation of neurotransmitter secretion
regulation of catalytic activity
response to stimulus
long-term synaptic potentiation
membrane fusion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
SH3 domain binding
Rab GTPase binding
GTPase regulator activity
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0