Search
Search

MLC1| Gene Symbol: MLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015166.3 | NP_055981.1 | 12 | 1854 | ||
| NM_139202.2 | NP_631941.1 | 12 | 1493 | |||
| XM_011530678.1 | XP_011528980.1 | 11 | 1450 | |||
| XM_017028671.1 | XP_016884160.1 | 11 | 1282 | |||
| GenBank mRNA | AF319633.1 | - | 12 | 1416 | ||
| AK124264.1 | - | 12 | 1855 | |||
| AK294048.1 | - | 10 | 1244 | |||
| AK295106.1 | - | 11 | 1115 | |||
| AK297341.1 | - | 11 | 1214 | |||
| AK299841.1 | - | 11 | 1262 | |||
| BC028425.2 | - | 12 | 1453 | 4817175 | ||
| BC070042.1 | - | 12 | 1451 | 5285102 | ||
| D25217.3 | - | 12 | 1530 | |||