Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIM2 SIRNA Assays ›
Gene Symbol:
TRIM2
Gene Name:
tripartite motif containing 2
Gene Aliases:
CMT2R, RNF86
Chromosome Location:
Chr.4: 153152281 - 153339322 on Build GRCh38
UniGene ID:
Hs.435711
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136859
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130067.1
NP_001123539.1
5
794
NM_015271.4
NP_056086.2
5
791
XM_006714157.1
XP_006714220.1
5
791
XM_006714158.1
XP_006714221.1
5
756
XM_006714159.3
XP_006714222.1
5
1602
XM_006714160.1
XP_006714223.1
6
1556
XM_006714161.1
XP_006714224.1
6
1553
XM_006714162.1
XP_006714225.1
6
1273
XM_006714165.3
XP_006714228.1
3
984
XM_011531794.1
XP_011530096.1
5
870
XM_011531795.1
XP_011530097.1
6
1276
XM_011531796.2
XP_011530098.1
5
806
XM_011531798.2
XP_011530100.1
5
911
XM_017007943.1
XP_016863432.1
5
756
XM_017007944.1
XP_016863433.1
6
1556
XM_017007945.1
XP_016863434.1
6
1553
XM_017007946.1
XP_016863435.1
5
1599
XM_017007947.1
XP_016863436.1
5
870
XM_017007948.1
XP_016863437.1
5
824
XM_017007949.1
XP_016863438.1
6
1273
XM_017007950.1
XP_016863439.1
6
1276
XM_017007951.1
XP_016863440.1
5
970
XM_017007952.1
XP_016863441.1
5
978
XM_017007953.1
XP_016863442.1
3
322
XM_017007954.1
XP_016863443.1
6
848
XM_017007955.1
XP_016863444.1
6
841
GenBank mRNA
AB011089.1
-
5
739
AF220018.1
-
5
739
AK308731.1
-
5
759
BC011052.1
-
5
794
4156234
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of neuron apoptotic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0