Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLASP1 SIRNA Assays ›
          Gene Symbol:
          CLASP1
          Gene Name:
          cytoplasmic linker associated protein 1
          Gene Aliases:
          MAST1
          Chromosome Location:
          Chr.2: 121337776 - 121649476 on Build GRCh38
          UniGene ID:
          Hs.469840
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136868

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CLASP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142273.1 NP_001135745.1 15, 16 1842
          NM_001142274.1 NP_001135746.1 15, 16 1842
          NM_001207051.1 NP_001193980.1 15, 16 1842
          NM_015282.2 NP_056097.1 15, 16 1842
          XM_006712377.1 XP_006712440.1 15, 16 1813
          XM_006712381.1 XP_006712444.1 15, 16 1813
          XM_006712382.1 XP_006712445.1 15, 16 1813
          XM_006712383.1 XP_006712446.1 15, 16 1813
          XM_006712385.1 XP_006712448.1 9, 10 885
          XM_011510848.1 XP_011509150.1 15, 16 1813
          XM_011510849.1 XP_011509151.1 15, 16 1813
          XM_017003657.1 XP_016859146.1 15, 16 1813
          XM_017003658.1 XP_016859147.1 15, 16 1813
          XM_017003659.1 XP_016859148.1 15, 16 1813
          XM_017003660.1 XP_016859149.1 15, 16 1813
          XM_017003661.1 XP_016859150.1 15, 16 1813
          XM_017003662.1 XP_016859151.1 15, 16 1813
          XM_017003663.1 XP_016859152.1 15, 16 1813
          XM_017003664.1 XP_016859153.1 15, 16 1813
          XM_017003665.1 XP_016859154.1 15, 16 1813
          XM_017003666.1 XP_016859155.1 15, 16 1813
          XM_017003667.1 XP_016859156.1 15, 16 1813
          XM_017003668.1 XP_016859157.1 15, 16 1813
          XM_017003669.1 XP_016859158.1 15, 16 1813
          XM_017003670.1 XP_016859159.1 15, 16 1813
          XM_017003671.1 XP_016859160.1 15, 16 1813
          XM_017003672.1 XP_016859161.1 15, 16 1813
          XM_017003673.1 XP_016859162.1 15, 16 1813
          XM_017003674.1 XP_016859163.1 15, 16 1813
          XM_017003675.1 XP_016859164.1 15, 16 1813
          XM_017003676.1 XP_016859165.1 8, 9 2647
          XM_017003677.1 XP_016859166.1 8, 9 2647
          XM_017003678.1 XP_016859167.1 8, 9 2646
          XM_017003679.1 XP_016859168.1 8, 9 2647
          XM_017003680.1 XP_016859169.1 8, 9 2647
          XM_017003681.1 XP_016859170.1 8, 9 2646
          XM_017003682.1 XP_016859171.1 8, 9 2646
          XM_017003683.1 XP_016859172.1 8, 9 2645
          XM_017003684.1 XP_016859173.1 8, 9 1628
          XM_017003685.1 XP_016859174.1 8, 9 2645
          XM_017003686.1 XP_016859175.1 8, 9 2645
          XM_017003687.1 XP_016859176.1 8, 9 1628
          XM_017003688.1 XP_016859177.1 8, 9 2645
          XM_017003689.1 XP_016859178.1 8, 9 1627
          XM_017003690.1 XP_016859179.1 15, 16 1813
          XM_017003691.1 XP_016859180.1 15, 16 1813
          GenBank mRNA AB014522.1 - 7, 8 705
          AF347693.1 - 14, 15 1617
          AK074338.1 - 2, 3 125
          AK091140.1 - 15, 16 1803
          AK297259.1 - 8, 9 796
          AK299331.1 - 10, 11 1364
          AK302393.1 - 9, 10 902
          BC112940.1 - 15, 16 1786 5501463
          BC132723.1 - 15, 16 1804 40146745
          BC144107.1 - 15, 16 1804 9052627
          CR933722.1 - 8, 9 794

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          microtubule cytoskeleton organization
          microtubule bundle formation
          vesicle targeting
          microtubule nucleation
          negative regulation of microtubule depolymerization
          Golgi organization
          mitotic spindle organization
          sister chromatid cohesion
          establishment or maintenance of cell polarity
          exit from mitosis
          regulation of gastrulation
          positive regulation of epithelial cell migration
          regulation of epithelial to mesenchymal transition
          astral microtubule organization
          microtubule organizing center organization
          negative regulation of microtubule polymerization or depolymerization
          positive regulation of microtubule polymerization
          microtubule anchoring
          establishment of mitotic spindle localization
          positive regulation of exocytosis
          establishment of spindle orientation
          cell division
          negative regulation of stress fiber assembly
          regulation of focal adhesion assembly
          regulation of microtubule cytoskeleton organization
          positive regulation of extracellular matrix disassembly
          establishment of epithelial cell polarity
          mitotic spindle assembly
          negative regulation of wound healing, spreading of epidermal cells
          positive regulation of basement membrane assembly involved in embryonic body morphogenesis
          dystroglycan binding
          protein binding
          microtubule binding
          kinetochore binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline