Search
Search

FAM175B| Gene Symbol: FAM175B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032182.3 | NP_115558.3 | 9 | 2487 | ||
| XM_011539554.1 | XP_011537856.1 | 8 | 2391 | |||
| XM_017015986.1 | XP_016871475.1 | 7 | 2365 | |||
| GenBank mRNA | BC008999.1 | - | 9 | 2449 | 2900632 | |
| D63877.1 | - | 9 | 2456 | |||