Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBADH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152740.3 | NP_689953.1 | 7 | 929 | ||
XM_017011719.1 | XP_016867208.1 | 8 | 959 | |||
GenBank mRNA | AB050000.1 | - | 7 | 803 | ||
AF529362.1 | - | 7 | 716 | |||
AK316605.1 | - | 7 | 734 | |||
BC013855.1 | - | 4 | 302 | |||
BC020167.1 | - | 7 | 790 | |||
BC032324.1 | - | 7 | 814 | 3890576 |