Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206957.1 | NP_001193886.1 | 3 | 503 | ||
NM_007182.4 | NP_009113.3 | 4 | 610 | |||
NM_170712.2 | NP_733830.1 | 4 | 320 | |||
NM_170713.2 | NP_733831.1 | 3 | 414 | |||
NM_170714.1 | NP_733832.1 | 4 | 622 | |||
XM_011533315.1 | XP_011531617.1 | 5 | 699 | |||
XM_011533316.2 | XP_011531618.1 | 3 | 518 | |||
GenBank mRNA | AF040703.2 | - | 3 | 327 | ||
AF061836.1 | - | 3 | 410 | |||
AF102770.2 | - | 4 | 515 | |||
AF102771.2 | - | 4 | 527 | |||
AF102772.2 | - | 4 | 527 | |||
AF132675.1 | - | 4 | 517 | |||
AF132676.1 | - | 3 | 280 | |||
AF132677.1 | - | 4 | 307 | |||
AF286217.1 | - | 3 | 408 | |||
AF291719.1 | - | 2 | 303 | |||
AK292638.1 | - | 3 | 404 | |||
BC110412.1 | - | 4 | 320 | 5756246 | ||
BC117153.1 | - | 4 | 586 | 40125704 | ||
BC143879.1 | - | 4 | 586 | 9052395 | ||
BI915594.1 | - | 3 | 425 | |||
BM562405.1 | - | 4 | 623 | |||
BQ072763.1 | - | 4 | 318 | |||
BT020047.1 | - | 3 | 269 | |||
BT020048.1 | - | 3 | 269 |