Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LDB2 SIRNA Assays ›
          Gene Symbol:
          LDB2
          Gene Name:
          LIM domain binding 2
          Gene Aliases:
          CLIM1, LDB-2, LDB1
          Chromosome Location:
          Chr.4: 16501534 - 16898809 on Build GRCh38
          UniGene ID:
          Hs.714330
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 137135

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LDB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130834.2 NP_001124306.1 5, 6 946
          NM_001290.4 NP_001281.1 5, 6 946
          NM_001304434.1 NP_001291363.1 5, 6 946
          NM_001304435.1 NP_001291364.1 5, 6 946
          XM_005248197.3 XP_005248254.1 5, 6 916
          XM_005248201.2 XP_005248258.1 5, 6 916
          XM_005248202.2 XP_005248259.1 5, 6 916
          XM_006713975.3 XP_006714038.1 7, 8 975
          XM_006713976.3 XP_006714039.1 7, 8 976
          XM_006713977.3 XP_006714040.1 7, 8 975
          XM_006713978.3 XP_006714041.1 7, 8 976
          XM_006713979.3 XP_006714042.1 7, 8 976
          XM_006713980.3 XP_006714043.1 7, 8 977
          XM_006713981.3 XP_006714044.1 7, 8 976
          XM_006713982.3 XP_006714045.1 7, 8 976
          XM_006713983.3 XP_006714046.1 7, 8 976
          XM_011513905.2 XP_011512207.1 6, 7 867
          XM_017008812.1 XP_016864301.1 7, 8 976
          XM_017008813.1 XP_016864302.1 5, 6 916
          XM_017008814.1 XP_016864303.1 7, 8 977
          XM_017008815.1 XP_016864304.1 7, 8 977
          XM_017008816.1 XP_016864305.1 7, 8 977
          XM_017008817.1 XP_016864306.1 5, 6 916
          XM_017008818.1 XP_016864307.1 5, 6 513
          XM_017008819.1 XP_016864308.1 5, 6 916
          XM_017008820.1 XP_016864309.1 5, 6 916
          XM_017008821.1 XP_016864310.1 5, 6 916
          XM_017008822.1 XP_016864311.1 6, 7 856
          GenBank mRNA AF047337.1 - 5, 6 731
          AF052389.1 - 6, 7 804
          AF064492.1 - 5, 6 731
          AF064493.1 - 5, 6 731
          AF068651.1 - 5, 6 729
          AK226029.1 - 5, 6 771
          AK294588.1 - 4, 5 501
          AK300119.1 - 5, 6 543
          AK312881.1 - 5, 6 641
          BC034019.1 - 5, 6 739 4693654

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          epithelial structure maintenance
          regulation of cell migration
          somatic stem cell population maintenance
          regulation of kinase activity
          positive regulation of cellular component biogenesis
          positive regulation of transcription from RNA polymerase II promoter
          transcription cofactor activity
          enzyme binding
          LIM domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0