Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304472.1 | NP_001291401.1 | 16 | 2201 | ||
NM_001304473.1 | NP_001291402.1 | 17 | 2227 | |||
NM_001304474.1 | NP_001291403.1 | 16 | 2172 | |||
NM_003741.3 | NP_003732.2 | 16 | 2201 | |||
NR_130747.1 | 14 | 2359 | ||||
XM_011513256.1 | XP_011511558.1 | 14 | 2210 | |||
XM_017007388.1 | XP_016862877.1 | 15 | 2135 | |||
XM_017007389.1 | XP_016862878.1 | 15 | 2135 | |||
XM_017007394.1 | XP_016862883.1 | 14 | 1923 | |||
GenBank mRNA | AB208947.1 | - | 10 | 3084 | ||
AF038198.1 | - | 3 | 305 | |||
AF076612.1 | - | 13 | 1493 | |||
AF209928.1 | - | 16 | 2201 | |||
AF209930.1 | - | 17 | 2227 | |||
AF283325.1 | - | 16 | 2172 | |||
AK074147.1 | - | 14 | 2112 | |||
AK293456.1 | - | 16 | 1923 | |||
AK300476.1 | - | 5 | 630 | |||
AY358926.1 | - | 16 | 2091 | |||
BC029879.1 | - | 3 | 331 | |||
BC112345.1 | - | 16 | 1986 | 40008417 |