Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003753.3 | NP_003744.1 | 14, 15 | 1808 | ||
GenBank mRNA | AK300199.1 | - | 14, 15 | 1678 | ||
AK301284.1 | - | 13, 14 | 1597 | |||
AK303884.1 | - | 14, 15 | 1650 | |||
BC000328.2 | - | 14, 15 | 1715 | 2822380 | ||
BC000469.2 | - | 14, 15 | 1727 | 2961048 | ||
BC014912.2 | - | 14, 15 | 1727 | 3915705 | ||
BC080515.1 | - | 14, 15 | 1714 | 2822489 | ||
BC093100.1 | - | 14, 15 | 1682 | 6527393 | ||
BC093686.1 | - | 14, 15 | 1670 | 7939531 | ||
BC101477.1 | - | 14, 15 | 1670 | 8068983 | ||
CR456489.1 | - | 14, 15 | 1727 | |||
U54558.1 | - | 14, 15 | 1727 |