Search
Search

TNFSF10| Gene Symbol: TNFSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190942.1 | NP_001177871.1 | 3 | 952 | ||
| NM_003810.3 | NP_003801.1 | 5 | 1100 | |||
| NR_033994.1 | 4 | 1057 | ||||
| GenBank mRNA | BC032722.1 | - | 5 | 1052 | 5504670 | |
| U37518.1 | - | 5 | 1064 | |||