Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RGN SIRNA Assays ›
          Gene Symbol:
          RGN
          Gene Name:
          regucalcin
          Gene Aliases:
          GNL, HEL-S-41, RC, SMP30
          Chromosome Location:
          Chr.X: 47078299 - 47093314 on Build GRCh38
          UniGene ID:
          Hs.77854
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 137586

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RGN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282848.1 NP_001269777.1 6 1613
          NM_001282849.1 NP_001269778.1 5 1446
          NM_004683.5 NP_004674.1 5 1042
          NM_152869.3 NP_690608.1 6 1662
          XM_006724567.2 XP_006724630.1 4 762
          GenBank mRNA AB028125.1 - 5 766
          AB032064.1 - 5 824
          AK223360.1 - 5 1042
          AK290136.1 - 5 980
          BC050371.1 - 5 1384 5760604
          BC073173.1 - 5 856 6186669
          D31815.1 - 5 765
          EU794655.1 - 5 824

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          negative regulation of protein kinase activity
          cellular calcium ion homeostasis
          spermatogenesis
          aging
          positive regulation of triglyceride biosynthetic process
          positive regulation of glucose metabolic process
          positive regulation of phosphatase activity
          L-ascorbic acid biosynthetic process
          negative regulation of phosphoprotein phosphatase activity
          positive regulation of ATPase activity
          negative regulation of GTPase activity
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          negative regulation of nitric oxide biosynthetic process
          positive regulation of fatty acid biosynthetic process
          negative regulation of epithelial cell proliferation
          regulation of calcium-mediated signaling
          negative regulation of cyclic-nucleotide phosphodiesterase activity
          liver regeneration
          negative regulation of sperm motility
          positive regulation of superoxide dismutase activity
          positive regulation of calcium-transporting ATPase activity
          negative regulation of RNA biosynthetic process
          negative regulation of bone development
          positive regulation of proteolysis involved in cellular protein catabolic process
          negative regulation of calcium-dependent ATPase activity
          negative regulation of DNA catabolic process
          positive regulation of dUTP diphosphatase activity
          negative regulation of leucine-tRNA ligase activity
          negative regulation of DNA biosynthetic process
          gluconolactonase activity
          calcium ion binding
          zinc ion binding
          enzyme regulator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0