Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004725.3 | NP_004716.1 | 8 | 1682 | ||
GenBank mRNA | AF047472.1 | - | 8 | 1510 | ||
AF053304.1 | - | 8 | 1478 | |||
AK226060.1 | - | 4256 | ||||
BX647392.1 | - | 7 | 2550 |