Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAGI1 SIRNA Assays ›
Gene Symbol:
MAGI1
Gene Name:
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases:
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location:
Chr.3: 65353525 - 66038836 on Build GRCh38
UniGene ID:
Hs.476636
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137663
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAGI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033057.1
NP_001028229.1
5
1333
NM_004742.2
NP_004733.2
5
1333
NM_015520.1
NP_056335.1
5
1333
XM_005265563.1
XP_005265620.1
5
1338
XM_005265564.1
XP_005265621.1
5
1335
XM_005265565.1
XP_005265622.1
5
1338
XM_005265566.1
XP_005265623.1
5
1335
XM_005265568.1
XP_005265625.1
5
1335
XM_005265570.1
XP_005265627.1
5
1338
XM_005265571.1
XP_005265628.1
5
1335
XM_005265574.3
XP_005265631.1
5
1335
XM_006713407.1
XP_006713470.1
5
1338
XM_006713408.1
XP_006713471.1
5
1335
XM_006713409.1
XP_006713472.1
5
1338
XM_006713410.1
XP_006713473.1
5
1335
XM_006713411.1
XP_006713474.1
5
1338
XM_006713412.1
XP_006713475.1
5
1335
XM_006713414.3
XP_006713477.1
5
1338
XM_011534236.1
XP_011532538.1
5
1338
XM_017007497.1
XP_016862986.1
5
1338
XM_017007498.1
XP_016862987.1
5
1335
XM_017007499.1
XP_016862988.1
5
1693
XM_017007500.1
XP_016862989.1
5
1689
XM_017007501.1
XP_016862990.1
5
1338
XM_017007502.1
XP_016862991.1
5
1338
XM_017007503.1
XP_016862992.1
5
1338
XM_017007504.1
XP_016862993.1
5
1335
XM_017007505.1
XP_016862994.1
5
1338
XM_017007506.1
XP_016862995.1
5
1335
XM_017007507.1
XP_016862996.1
4
499
XM_017007508.1
XP_016862997.1
4
499
GenBank mRNA
AB010894.1
-
5
1049
AF401654.1
-
5
807
AF401655.1
-
5
807
AF401656.1
-
5
807
AK289803.1
-
5
902
AK310365.1
-
10
1334
BC062367.1
-
5
1336
KC170987.1
-
5
807
U80754.1
-
3
360
U96115.1
-
3
354
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cell adhesion
cell surface receptor signaling pathway
neuron death
protein binding
ATP binding
protein C-terminus binding
protein complex scaffold
alpha-actinin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8j2zf:80/100.66.76.51:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0