Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004869.3 | NP_004860.2 | 7 | 1016 | ||
XM_006722582.2 | XP_006722645.1 | 6 | 892 | |||
XM_011526268.2 | XP_011524570.1 | 7 | 1016 | |||
GenBank mRNA | AF038960.1 | - | 7 | 945 | ||
AF195514.1 | - | 7 | 957 | |||
AK290932.1 | - | 7 | 960 | |||
AK291253.1 | - | 7 | 914 | |||
AK308892.1 | - | 7 | 980 | |||
AK309322.1 | - | 6 | 892 | |||
AK311041.1 | - | 6 | 892 | |||
AY232629.1 | - | 7 | 766 | |||
BC039574.1 | - | 7 | 923 | 6042862 |