Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127393.1 | NP_001120865.1 | 5 | 787 | ||
NM_004894.2 | NP_004885.1 | 4 | 518 | |||
GenBank mRNA | AF054175.1 | - | 4 | 481 | ||
BC000429.2 | - | 4 | 470 | 2819801 | ||
BC001944.1 | - | 4 | 470 | 2819801 | ||
BX161388.1 | - | 5 | 722 |