Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SQSTM1 SIRNA Assays ›
          Gene Symbol:
          SQSTM1
          Gene Name:
          sequestosome 1
          Gene Aliases:
          A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B
          Chromosome Location:
          Chr.5: 179806388 - 179838078 on Build GRCh38
          UniGene ID:
          Hs.587290
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 13813

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SQSTM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142298.1 NP_001135770.1 3 359
          NM_001142299.1 NP_001135771.1 3 276
          NM_003900.4 NP_003891.1 2 346
          XM_017010010.1 XP_016865499.1 2 578
          GenBank mRNA AK025146.1 - 2 65
          AK098077.1 - 1 519
          AK226167.1 - 2 68
          AK293509.1 - 2 283
          AK304877.1 - 2 283
          AK312451.1 - 2 310
          BC000951.2 - 1 35
          BC001874.1 - 4 510 3535436
          BC003139.1 - 2 269 2906264
          BC017222.1 - 2 291 4298142
          BC019111.1 - 2 283 5091391
          BI549805.1 - 3 364
          BM800056.1 - 297
          DA541885.1 - 3 276
          DC398116.1 - 2 346
          U41806.1 - 2 271
          U46751.1 - 2 303

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy
          positive regulation of protein phosphorylation
          immune system process
          protein phosphorylation
          ubiquitin-dependent protein catabolic process
          autophagy
          apoptotic process
          response to stress
          protein localization
          regulation of mitochondrion organization
          endosomal transport
          macroautophagy
          positive regulation of macroautophagy
          cell differentiation
          intracellular signal transduction
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of growth of symbiont in host
          positive regulation of transcription from RNA polymerase II promoter
          regulation of Ras protein signal transduction
          protein heterooligomerization
          regulation of protein complex stability
          mitophagy in response to mitochondrial depolarization
          protein serine/threonine kinase activity
          protein kinase C binding
          protein binding
          zinc ion binding
          protein kinase binding
          receptor tyrosine kinase binding
          ubiquitin protein ligase binding
          SH2 domain binding
          identical protein binding
          protein homodimerization activity
          ubiquitin binding
          K63-linked polyubiquitin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0