Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280558.1 | NP_001267487.1 | 6 | 772 | ||
NM_006032.3 | NP_006023.1 | 5 | 760 | |||
XM_005268216.1 | XP_005268273.1 | 6 | 716 | |||
XM_005268217.1 | XP_005268274.1 | 4 | 410 | |||
GenBank mRNA | AB009288.1 | - | 5 | 560 | ||
AK125195.1 | - | 4 | 1322 | |||
AK222675.1 | - | 5 | 520 | |||
AK293655.1 | - | 6 | 772 | |||
AK294460.1 | - | 4 | 675 | |||
AK314182.1 | - | 5 | 573 | |||
AL137688.1 | - | 3 | 1971 | |||
BC018046.1 | - | 5 | 566 | 4799650 |