Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PICK1 SIRNA Assays ›
          Gene Symbol:
          PICK1
          Gene Name:
          protein interacting with PRKCA 1
          Gene Aliases:
          PICK, PRKCABP
          Chromosome Location:
          Chr.22: 38056311 - 38075704 on Build GRCh38
          UniGene ID:
          Hs.180871
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138239
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PICK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039583.1 NP_001034672.1 7, 8 783
          NM_001039584.1 NP_001034673.1 7, 8 730
          NM_012407.3 NP_036539.1 7, 8 881
          XM_006724379.2 XP_006724442.1 6, 7 667
          XM_006724380.2 XP_006724443.1 6, 7 586
          XM_017029090.1 XP_016884579.1 7, 8 845
          XM_017029091.1 XP_016884580.1 7, 8 896
          XM_017029092.1 XP_016884581.1 7, 8 808
          XM_017029093.1 XP_016884582.1 5, 6 553
          XM_017029094.1 XP_016884583.1 6, 7 859
          XM_017029095.1 XP_016884584.1 9, 10 1472
          GenBank mRNA AB026491.1 - 7, 8 694
          AF231710.1 - 6, 7 503
          AK000875.1 - 7, 8 637
          AK092818.1 - 7, 8 731
          AK222640.1 - 7, 8 729
          AK299684.1 - 7, 8 578
          AK308792.1 - 6, 7 488
          AK310629.1 - 8, 9 979
          AL049654.1 - 7, 8 718
          BC017561.2 - 7, 8 699 4026028
          CR456550.1 - 7, 8 604

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00202661_m1

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of receptor internalization
          protein phosphorylation
          retrograde vesicle-mediated transport, Golgi to ER
          protein kinase C-activating G-protein coupled receptor signaling pathway
          monoamine transport
          glial cell development
          regulation of Arp2/3 complex-mediated actin nucleation
          negative regulation of Arp2/3 complex-mediated actin nucleation
          cellular response to decreased oxygen levels
          cellular response to glucose starvation
          DNA methylation involved in embryo development
          DNA methylation involved in gamete generation
          receptor clustering
          neuronal ion channel clustering
          long term synaptic depression
          dendritic spine organization
          dendritic spine maintenance
          G-protein coupled receptor binding
          protein kinase C binding
          receptor binding
          protein binding
          protein C-terminus binding
          ATPase activity
          enzyme binding
          protein domain specific binding
          identical protein binding
          metal ion binding
          actin filament binding
          Arp2/3 complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0