Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZEB2 SIRNA Assays ›
          Gene Symbol:
          ZEB2
          Gene Name:
          zinc finger E-box binding homeobox 2
          Gene Aliases:
          HSPC082, SIP-1, SIP1, SMADIP1, ZFHX1B
          Chromosome Location:
          Chr.2: 144384375 - 144520391 on Build GRCh38
          UniGene ID:
          Hs.34871
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138439

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZEB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171653.1 NP_001165124.1 7 1769
          NM_014795.3 NP_055610.1 8 1841
          XM_006712881.3 XP_006712944.1 8 1435
          XM_006712882.3 XP_006712945.1 8 1484
          XM_011512232.2 XP_011510534.1 7 1478
          XM_017005414.1 XP_016860903.1 9 1555
          XM_017005415.1 XP_016860904.1 7 2458
          GenBank mRNA AB011141.1 - 8 1763
          AB056507.1 - 8 1798
          AB193095.1 - 1699
          AB193096.1 - 7 1319
          AK294928.1 - 7 1497
          AK308445.1 - 8 1448
          BC127101.1 - 8 1453 40124119
          BC127102.1 - 8 1452 40124124

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_015753, NM_001289521, BC060699, AK141157, AK122312, and others.

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural crest cell migration
          somitogenesis
          neural tube closure
          transcription from RNA polymerase II promoter
          nervous system development
          corpus callosum morphogenesis
          hippocampus development
          cell proliferation in forebrain
          corticospinal tract morphogenesis
          positive regulation of Wnt signaling pathway
          positive regulation of JUN kinase activity
          positive regulation of melanocyte differentiation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of melanin biosynthetic process
          developmental pigmentation
          collateral sprouting
          positive regulation of axonogenesis
          mammillary axonal complex development
          melanocyte migration
          positive regulation of lens fiber cell differentiation
          regulation of melanosome organization
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          protein binding
          phosphatase regulator activity
          metal ion binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0