Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099691.2 | NP_001093161.1 | 6 | 1590 | ||
NM_001308158.1 | NP_001295087.1 | 6 | 1760 | |||
NM_001308159.1 | NP_001295088.1 | 6 | 1757 | |||
NM_003236.3 | NP_003227.1 | 6 | 1593 | |||
GenBank mRNA | CN274505.1 | - | 1 | 340 | ||
X70340.1 | - | 6 | 1376 |