Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTC3 SIRNA Assays ›
Gene Symbol:
TTC3
Gene Name:
tetratricopeptide repeat domain 3
Gene Aliases:
DCRR1, RNF105, TPRDIII
Chromosome Location:
Chr.21: 37073184 - 37203118 on Build GRCh38
UniGene ID:
Hs.368214
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138943
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTC3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001894.1
NP_001001894.1
12
1115
NM_001320703.1
NP_001307632.1
12
1321
NM_001320704.1
NP_001307633.1
12
1311
NM_003316.3
NP_003307.3
12
2406
XM_005261048.1
XP_005261105.1
13
1221
XM_005261049.1
XP_005261106.1
13
1144
XM_005261050.4
XP_005261107.1
13
4106
XM_005261052.1
XP_005261109.1
12
1096
XM_005261054.3
XP_005261111.1
12
4050
XM_005261055.1
XP_005261112.1
3
276
XM_005261056.2
XP_005261113.1
3
355
XM_011529739.2
XP_011528041.1
5
427
XM_011529740.2
XP_011528042.1
10
935
XM_011529742.2
XP_011528044.2
13
1166
XM_017028458.1
XP_016883947.1
13
1160
XM_017028459.1
XP_016883948.1
13
1159
XM_017028460.1
XP_016883949.1
12
1110
XM_017028461.1
XP_016883950.1
13
1221
XM_017028462.1
XP_016883951.1
13
1144
XM_017028463.1
XP_016883952.1
12
1173
XM_017028464.1
XP_016883953.1
3
395
XM_017028465.1
XP_016883954.1
9
2506
XM_017028466.1
XP_016883955.1
3
371
XM_017028467.1
XP_016883956.1
3
276
GenBank mRNA
AJ001866.1
-
9
1167
AK125473.1
-
12
3758
AK291992.1
-
12
1223
AK297921.1
-
4
402
AK299991.1
-
3
252
BC047867.1
-
6
507
BC063033.1
-
12
1133
BC092466.1
-
12
1118
BC137345.1
-
12
1545
9021343
BI915053.1
-
3
292
CT002499.1
-
5
410
D83077.1
-
12
2406
D83327.1
-
9
758
D84294.1
-
11
2479
D84295.1
-
10
2393
D84296.1
-
2
817
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108315, XM_008768582.This siRNA also targets the following transcripts in Mouse: NM_009441, BC057207, AK220495, AK166164, AB008516, and others.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
ubiquitin-dependent protein catabolic process
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0