Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST2H2BE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003528.2 | NP_003519.1 | 1 | 1256 | ||
GenBank mRNA | AK093747.1 | - | 2 | 1216 | ||
BC005827.2 | - | 1 | 1226 | 2989788 | ||
BC069193.1 | - | 1 | 1238 | 3936695 | ||
M60756.1 | - | 1 | 1135 |