Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PKP4 SIRNA Assays ›
Gene Symbol:
PKP4
Gene Name:
plakophilin 4
Gene Aliases:
p0071
Chromosome Location:
Chr.2: 158456880 - 158681429 on Build GRCh38
UniGene ID:
Hs.407580
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139244
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PKP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005476.2
NP_001005476.1
5
654
NM_001304969.1
NP_001291898.1
5
654
NM_001304970.1
NP_001291899.1
5
654
NM_001304971.1
NP_001291900.1
5
654
NM_003628.4
NP_003619.2
5
654
XM_011512016.1
XP_011510318.1
5
1091
XM_011512017.1
XP_011510319.1
5
401
XM_011512018.1
XP_011510320.1
5
1091
XM_011512019.1
XP_011510321.1
5
401
XM_011512020.2
XP_011510322.1
5
438
XM_011512021.2
XP_011510323.1
5
438
XM_011512022.1
XP_011510324.1
4
606
XM_011512026.2
XP_011510328.1
5
438
XM_017005122.1
XP_016860611.1
5
1091
XM_017005123.1
XP_016860612.1
5
1626
XM_017005124.1
XP_016860613.1
5
1091
XM_017005125.1
XP_016860614.1
5
438
XM_017005126.1
XP_016860615.1
5
401
XM_017005127.1
XP_016860616.1
5
1091
XM_017005128.1
XP_016860617.1
5
438
XM_017005129.1
XP_016860618.1
4
606
XM_017005130.1
XP_016860619.1
4
606
XM_017005132.1
XP_016860621.1
3
178
XM_017005133.1
XP_016860622.1
5
608
XM_017005134.1
XP_016860623.1
4
978
XM_017005135.1
XP_016860624.1
3
178
GenBank mRNA
AK054911.1
-
5
1168
AK055823.1
-
746
AK128720.1
-
3
487
BC048013.1
-
5
570
BC050308.1
-
5
436
5262792
X81889.1
-
5
451
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_003749476, XM_006234220, XM_003749475, XM_006234216, XM_006234218, and others.
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cell-cell junction assembly
cell-cell signaling
single organismal cell-cell adhesion
regulation of cell adhesion
positive regulation of cytokinesis
positive regulation of GTPase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0