Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MADD SIRNA Assays ›
          Gene Symbol:
          MADD
          Gene Name:
          MAP kinase activating death domain
          Gene Aliases:
          DENN, IG20, RAB3GEP
          Chromosome Location:
          Chr.11: 47269376 - 47330031 on Build GRCh38
          UniGene ID:
          Hs.82548
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 139310

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MADD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135943.1 NP_001129415.1 9, 10 1915
          NM_001135944.1 NP_001129416.1 9, 10 1891
          NM_003682.3 NP_003673.3 9, 10 1891
          NM_130470.2 NP_569826.2 9, 10 2057
          NM_130471.2 NP_569827.2 9, 10 1891
          NM_130472.2 NP_569828.2 9, 10 1891
          NM_130473.2 NP_569829.2 9, 10 1891
          NM_130474.2 NP_569830.2 9, 10 1891
          NM_130475.2 NP_569831.1 9, 10 1915
          NM_130476.2 NP_569832.2 9, 10 1915
          XM_005253189.2 XP_005253246.1 9, 10 1915
          XM_005253196.2 XP_005253253.1 9, 10 1915
          XM_005253199.2 XP_005253256.1 9, 10 1915
          XM_005253200.2 XP_005253257.1 9, 10 1915
          XM_005253201.2 XP_005253258.1 9, 10 1915
          XM_005253203.2 XP_005253260.1 9, 10 1915
          XM_005253204.2 XP_005253261.1 9, 10 1915
          XM_005253205.1 XP_005253262.1 9, 10 1915
          XM_017018478.1 XP_016873967.1 9, 10 1915
          XM_017018479.1 XP_016873968.1 9, 10 1915
          XM_017018480.1 XP_016873969.1 9, 10 1915
          XM_017018481.1 XP_016873970.1 9, 10 1915
          XM_017018482.1 XP_016873971.1 9, 10 1915
          XM_017018483.1 XP_016873972.1 9, 10 1915
          XM_017018484.1 XP_016873973.1 9, 10 1915
          XM_017018485.1 XP_016873974.1 9, 10 1876
          XM_017018486.1 XP_016873975.1 9, 10 1915
          XM_017018487.1 XP_016873976.1 9, 10 1915
          XM_017018488.1 XP_016873977.1 9, 10 1915
          XM_017018489.1 XP_016873978.1 9, 10 1876
          XM_017018490.1 XP_016873979.1 9, 10 1902
          XM_017018491.1 XP_016873980.1 9, 10 1871
          XM_017018492.1 XP_016873981.1 9, 10 1915
          XM_017018493.1 XP_016873982.1 9, 10 1902
          XM_017018494.1 XP_016873983.1 9, 10 1915
          XM_017018495.1 XP_016873984.1 9, 10 1915
          XM_017018496.1 XP_016873985.1 9, 10 1871
          XM_017018497.1 XP_016873986.1 9, 10 1876
          XM_017018498.1 XP_016873987.1 9, 10 1915
          XM_017018499.1 XP_016873988.1 9, 10 1884
          XM_017018500.1 XP_016873989.1 9, 10 1915
          XM_017018501.1 XP_016873990.1 9, 10 1871
          XM_017018502.1 XP_016873991.1 9, 10 1884
          XM_017018503.1 XP_016873992.1 9, 10 1871
          XM_017018504.1 XP_016873993.1 9, 10 1884
          XM_017018505.1 XP_016873994.1 9, 10 1871
          XM_017018506.1 XP_016873995.1 9, 10 1876
          XM_017018507.1 XP_016873996.1 9, 10 1902
          XM_017018508.1 XP_016873997.1 9, 10 1915
          XM_017018509.1 XP_016873998.1 9, 10 1871
          XM_017018510.1 XP_016873999.1 9, 10 1915
          GenBank mRNA AB002356.1 - 9, 10 1887
          AF440100.1 - 9, 10 1865
          AF440101.1 - 9, 10 1865
          AF440102.1 - 9, 10 1865
          AF440103.1 - 9, 10 1865
          AF440434.1 - 9, 10 1865
          AK292442.1 - 9, 10 1855
          BC040484.1 - 9, 10 1899 5271798
          U44953.1 - 9, 10 1879
          U77352.1 - 9, 10 2025

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          cell surface receptor signaling pathway
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of Rab protein signal transduction
          regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of cell cycle
          execution phase of apoptosis
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of extrinsic apoptotic signaling pathway
          death receptor binding
          protein binding
          Rab guanyl-nucleotide exchange factor activity
          protein kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0