Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004615.3 | NP_004606.2 | 5 | 608 | ||
GenBank mRNA | AB062057.1 | - | 5 | 609 | ||
AB102665.1 | - | 5 | 524 | |||
AK293178.1 | - | 6 | 593 | |||
AK293534.1 | - | 7 | 762 | |||
AK293552.1 | - | 6 | 673 | |||
AK312343.1 | - | 5 | 542 | |||
BC018036.1 | - | 5 | 567 | 4801388 | ||
D10653.1 | - | 5 | 548 | |||
D29808.1 | - | 5 | 534 | |||
L10373.1 | - | 5 | 592 |