Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FXR1 SIRNA Assays ›
          Gene Symbol:
          FXR1
          Gene Name:
          FMR1 autosomal homolog 1
          Gene Aliases:
          FXR1P
          Chromosome Location:
          Chr.3: 180912446 - 180982753 on Build GRCh38
          UniGene ID:
          Hs.478407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 139486

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FXR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013438.2 NP_001013456.1 5, 6 665
          NM_001013439.2 NP_001013457.1 6, 7 1009
          NM_005087.3 NP_005078.2 5, 6 665
          XM_005247813.3 XP_005247870.2 5, 6 440
          XM_005247814.3 XP_005247871.2 5, 6 440
          XM_005247815.3 XP_005247872.2 5, 6 440
          XM_005247816.3 XP_005247873.2 5, 6 440
          XM_006713775.3 XP_006713838.1 6, 7 783
          XM_017007298.1 XP_016862787.1 6, 7 783
          XM_017007299.1 XP_016862788.1 6, 7 783
          XM_017007300.1 XP_016862789.1 6, 7 783
          XM_017007301.1 XP_016862790.1 6, 7 783
          GenBank mRNA AK225334.1 - 5, 6 441
          AK292633.1 - 5, 6 441
          AK294603.1 - 4, 5 387
          AK297254.1 - 3, 4 293
          AK297331.1 - 4, 5 346
          AK302197.1 - 5, 6 444
          AY341428.1 - 5, 6 716
          BC028983.1 - 5, 6 435 5122574
          BX956500.1 - 5, 6 451
          KR259877.1 - 5, 6 425
          KR259878.1 - 5, 6 425
          KR259879.1 - 5, 6 425
          KR259880.1 - 6, 7 769
          U25165.1 - 5, 6 437

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          muscle organ development
          negative regulation of translation
          cell differentiation
          positive regulation of gene silencing by miRNA
          G-quadruplex RNA binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          protein binding
          RNA strand annealing activity
          protein homodimerization activity
          poly(A) RNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0