Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005654.5 | NP_005645.1 | 3 | 3187 | ||
XM_017009797.1 | XP_016865286.1 | 3 | 1180 | |||
GenBank mRNA | AF087978.1 | - | 1 | 54 | ||
BC004154.2 | - | 3 | 1588 | 2824138 | ||
BC017493.1 | - | 3 | 3187 | 3912075 | ||
X16155.1 | - | 3 | 1485 |