Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320952.1 | NP_001307881.1 | 2 | 670 | ||
NM_001320953.1 | NP_001307882.1 | 4 | 957 | |||
NM_001320954.1 | NP_001307883.1 | 4 | 902 | |||
NM_001320955.1 | NP_001307884.1 | 2 | 670 | |||
NM_014519.3 | NP_055334.2 | 2 | 670 | |||
XM_011524006.2 | XP_011522308.1 | 3 | 336 | |||
XM_017025021.1 | XP_016880510.1 | 3 | 887 | |||
XM_017025022.1 | XP_016880511.1 | 3 | 337 | |||
XM_017025023.1 | XP_016880512.1 | 3 | 338 | |||
GenBank mRNA | AK057506.1 | - | 2 | 360 | ||
AK225622.1 | - | 2 | 227 | |||
AK297835.1 | - | 2 | 272 | |||
AK298190.1 | - | 3 | 419 | |||
AK316369.1 | - | 4 | 1031 | |||
BC002852.1 | - | 246 | ||||
BC016039.1 | - | 2 | 255 | 4635999 | ||
Y15067.1 | - | 2 | 262 |