Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014565.2 | NP_055380.2 | 1 | 796 | ||
GenBank mRNA | BC069169.1 | - | 1 | 796 | 7216916 | |
BC128248.1 | - | 1 | 796 | |||
BC141934.1 | - | 1 | 796 | 40114773 | ||
BC141935.1 | - | 1 | 796 | 40114774 |