Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STAU1 SIRNA Assays ›
Gene Symbol:
STAU1
Gene Name:
staufen double-stranded RNA binding protein 1
Gene Aliases:
PPP1R150, STAU
Chromosome Location:
Chr.20: 49113339 - 49219295 on Build GRCh38
UniGene ID:
Hs.596704
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139816
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STAU1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037328.2
NP_001032405.1
4
561
NM_001319134.1
NP_001306063.1
3
438
NM_001319135.1
NP_001306064.1
5
638
NM_001322927.1
NP_001309856.1
2
363
NM_001322928.1
NP_001309857.1
5
707
NM_001322929.1
NP_001309858.1
5
850
NM_001322930.1
NP_001309859.1
3
652
NM_001322931.1
NP_001309860.1
6
830
NM_001322932.1
NP_001309861.1
3
652
NM_001322933.1
NP_001309862.1
4
727
NM_004602.3
NP_004593.2
4
561
NM_017452.3
NP_059346.2
3
438
NM_017453.3
NP_059347.2
4
727
NM_017454.3
NP_059348.2
2
363
XM_005260527.1
XP_005260584.1
4
610
XM_006723865.1
XP_006723928.1
5
847
XM_011529015.1
XP_011527317.1
6
960
XM_017028028.1
XP_016883517.1
5
704
GenBank mRNA
AB209561.1
-
4
592
AF061938.1
-
3
432
AF061939.1
-
4
721
AF061940.1
-
2
357
AF061941.1
-
4
564
AJ132258.1
-
1
105
AK025519.1
-
3
401
AK057739.1
-
4
684
AK291487.1
-
4
642
AK292859.1
-
3
223
AL136601.2
-
3
421
AY529074.1
-
3
256
AY546099.1
-
3
432
BC001893.1
-
4
365
BC010169.2
-
4
365
BC050432.1
-
3
465
6047265
BC095397.1
-
3
410
30528051
DA455473.1
-
3
437
DA858410.1
-
3
436
DB086071.1
-
4
350
DB100321.1
-
3
437
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to oxidative stress
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
positive regulation of long-term synaptic potentiation
double-stranded RNA binding
protein binding
protein phosphatase 1 binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-6dgdk:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0